Please login to your online account to display your discounted pricing

Alfa Aesar™ CTX IV (6-12)

For cellular and molecular biology applications

Specifications

Product Type CTX IV (6-12)
Molecular Weight 899.15
Storage Requirements -30° to -10°C
Sequence Leu-Ile-Pro-Pro-Phe-Trp-Lys-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66101LB0

 
alfa aesar™
J66101LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66101MA

 
alfa aesar™
J66101MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type CTX IV (6-12)
Molecular Weight 899.15
Storage Requirements -30° to -10°C
Sequence Leu-Ile-Pro-Pro-Phe-Trp-Lys-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC