missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ CTAP

For cellular and molecular biology applications

$221.39 - $406.16

Specifications

Product Type CTAP
Molecular Weight 1104.30
Storage Requirements -30° to -10°C
Sequence D-Phe-Cys-Tyr-D-Trp-Arg-Thr-Pen-Thr-NH2 (Disulfide bridge: Cys2-Pen7)
Format Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66219MCR View Documents Alfa Aesar™
J66219MCR
1mg Each for $221.39
 
AAJ66219EXC View Documents Alfa Aesar™
J66219EXC
2.5mg Each for $406.16
 
Specifications

Specifications

CTAP
-30° to -10°C
Solid
HPLC
1104.30
D-Phe-Cys-Tyr-D-Trp-Arg-Thr-Pen-Thr-NH2 (Disulfide bridge: Cys2-Pen7)
RUO

For Research Use Only