Invitrogen™ CSNK2A2 (CK2 α 2) Recombinant Human Protein DFS Item
Casein kinase II is a protein serine/threonine kinase that is thought to be present in all eukarotic cells, implying that it has fundamental cellular functions.
Manufacturer:  Invitrogen™ LSPV3993
 View more versions of this product
Catalog No. PV3993
Description
Description
The CSNK2A2 holoenzyme is a tetramer containing 2 alpha or alpha-prime subunits (or one of each) and 2 beta subunits. The function of beta subunits is unknown and the alpha subunit is the catalytic unit.
Specifications
Specifications
NP_001887
Frozen
69.2kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
CK2 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
CSNK2A2 (CK2 α 2)
Baculovirus
Human
GST Tag
Documents