Invitrogen™ CSNK1G1 (CK1 γ 1) Recombinant Human Protein DFS Item
CSNK1G1 (CK1 gamma 1) is one of the most abundant serine/threonine kinases in eukaryotic cell extracts.
Manufacturer:  Invitrogen™ LSPV3989
 View more versions of this product
Catalog No. PV3989
Description
Description
Multiple isoforms of the CSNK1 enzyme exist, and the gamma-1 isoform is involved in growth and morphogenesis of eukaryotic cells.
Specifications
Specifications
NP_071331
Frozen
72.9kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
CK1 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
CSNK1G1 (CK1 γ 1)
Baculovirus
Human
GST Tag
Documents