Invitrogen™ CSNK1D (CK1 δ) Recombinant Human Protein DFS Item
CSNK1D (CK1 delta), a serine/threonine kinase, plays a critical role in Wnt/beta-catenin signaling pathway.
Manufacturer:  Invitrogen™ LSPV3987
 View more versions of this product
Catalog No. PV3987
Description
Description
The Wnt/beta-catenin signaling pathway is important in both development and cancer.
Specifications
Specifications
NP_620693
Frozen
74.3kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
CK1 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
0mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
CSNK1D (CK1 δ)
Baculovirus
Human
GST Tag
Documents