missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Human CSNK1A1 (CK1 alpha 1), GST Tag Recombinant Protein
Human CSNK1A1 (CK1 alpha 1), GST Tag Recombinant Protein
$2645.00 - $11260.00
Specifications
Accession Number | NP_001883, K164Q |
---|---|
Gene Alias | CSNK1A1 (CK1 α 1), CK1 |
KinaseFamily | CK1 Kinase Family |
Species | Human |
Recombinant | Recombinant |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR7361B
|
Invitrogen™
PR7361B |
100μg |
Each for $2,645.00
|
|
|||||
PV4174
|
Invitrogen™
PV4174 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
CKI proteins are monomeric, ranging from 25 to 55 kD, and are found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cells.
Specifications
NP_001883, K164Q | |
CK1 Kinase Family | |
Recombinant | |
Baculovirus |
CSNK1A1 (CK1 α 1), CK1 | |
Human | |
GST Tag |