Invitrogen™ CSNK1A1 (CK1 α 1) Recombinant Human Protein DFS Item
CNSK1A1 (CK1 alpha 1) is a member of the casein kinase I (CKI) gene family, which encodes serine/threonine kinases that preferentially phosphorylate acidic substrates.
Manufacturer:  Invitrogen™ LSPV4174
 View more versions of this product
Catalog No. PV4174
Description
Description
CKI proteins are monomeric, ranging from 25 to 55 kD, and are found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cells.
Specifications
Specifications
NP_001883, K164Q
Frozen
66.4kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
CK1 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
CSNK1A1 (CK1 α 1), CK1
Baculovirus
Human
GST Tag
Documents