CSK Recombinant Human Protein DFS Item
CSK Recombinant Human Protein
Manufacturer:  Life Technologies LSP2927
 View more versions of this product
Catalog No. P2927
Description
Description
CSK is a tyrosine kinase that has been shown to downregulate the activity of SRC oncoprotein through phosphorylation of its carboxy terminus. Since cell transformation by SRC is caused by various mechanisms that interfere with this phosphorylation, the CSK gene might function as an antioncogene.
Specifications
Specifications
NP_004374
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
51.9kDa
95% by SDS-PAGE
–80°C
1 Each
CSK
CSK was pre-diluted in enzyme dilution buffer and assayed in 60mM Tris (pH 7.5), 4mM MnCl2, 0.05% NP-40, 200μM ATP, 500μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
CSK
1110nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL
His Tag
0.34mg/mL
1445
CSK Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Cytoplasmic tyrosine kinase
CSK Recombinant Human Protein
Kinase assay
E. coli
CSK Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.