Crystalgen Bio-Degradable Green Culture Flasks
Manufacturer:  Crystalgen 260005
 View more versions of this product
Catalog No. 05-539-808
Description
Description
100% bio-degradable lab disposable plastic culture flasks
  • Made of raw polymer that can be degraded by the soil within a year after disposal or 70 days under composting conditions
  • Certified by LNE EN13432 biodegradable report and FDA Biomedical Applications
Available in Standard or Baffled
All shaking cultures, mixing, and storage of biomedical fluids
Specifications
Specifications
Baffled
125 mL
Yes
4.22 oz.
Plastic
For use with all shaking cultures, mixing and storage of biomedical fluids, it comes from nature and is renewable
Documents