Please login to your online account to display your discounted pricing

Crystal Violet, HARLECO™, EMD Millipore


Quantity 25g

Manufacturer: emd millipore corporation  19212

 View more versions of this product

Catalog No. M19212

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 25g
Color Violet
Purity Grade Certified Biological Stain
Physical Form Powder
Chemical Name or Material Crystal Violet
Name Note HARLECO™
Molecular Formula C25N3H30Cl
CAS 467-63-0
Synonym Gentian Violet

Specifications

Chemical Name or Material Crystal Violet
Name Note HARLECO™
CAS 467-63-0
Purity Grade Certified Biological Stain
Physical Form Powder
Molecular Formula C25N3H30Cl
Synonym Gentian Violet