Please login to your online account to display your discounted pricing

Crystal Violet, HARLECO™, EMD Millipore

$155.46 - $439.09

Chemical Identifiers

Chemical Name or Material Crystal Violet
Name Note HARLECO™
CAS 467-63-0
Purity Grade Certified Biological Stain
Physical Form Powder
Molecular Formula C25N3H30Cl
Synonym Gentian Violet
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

M19216

 
emd millipore corporation
19216
100g Each for $439.09

M19212

 
emd millipore corporation
19212
25g Each for $155.46
Description & Specifications

Specifications

Quantity 100g
Color Violet
Purity Grade Certified Biological Stain
Physical Form Powder
Chemical Name or Material Crystal Violet
Name Note HARLECO™
Molecular Formula C25N3H30Cl
CAS 467-63-0
Synonym Gentian Violet