Please login to your online account to display your discounted pricing

Crystal Violet, 1% (w/v) in Glacial Acetic Acid, for Non-Aqueous Titrations, Certified, LabChem™

$54.26 - $54.26

Chemical Identifiers

Chemical Name or Material Crystal Violet
CAS 64-19-7
CAS Max % 99
CAS 548-62-9
CAS Max % 1
Name Note 1% (w/v) in Glacial Acetic Acid, for Non-Aqueous Titrations
Purity Grade Certified Grade
Linear Formula C25H30CIN3
Formula Weight 407.99
UN Number UN2789
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC135417

 
lab chem
LC135417
Amber Glass 125mL Each for $54.26
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 407.99
Chemical Name or Material Crystal Violet
Quantity 125mL
Flash Point 40°C
Name Note 1% (w/v) in Glacial Acetic Acid, for Non-Aqueous Titrations
Physical Form Liquid
Linear Formula C25H30CIN3
CAS 64-19-7
UN Number UN2789
Identification Passes Test
Health Hazard 1 Danger
Odor Vinegar
Packaging Amber Glass
CAS Max % 99
Health Hazard 2 GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Harmful to aquatic life with long lasting effects.
Viscosity 2.18cSt
CAS 548-62-9
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhahled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Specific Gravity 1.06g/mL
Suitability Passes Test
CAS Max % 1
Recommended Storage Room Temperature
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases; Thermal decomposition generates: Corrosive vapors
Density 1.06g/mL
Boiling Point 118°C
Color Dark purple
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhahled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Harmful to aquatic life with long lasting effects.

Room Temperature