Crystal Models
Durable, permanently bonded crystalline models.
Manufacturer:  Fisher Scientific FOM530
 View more versions of this product
Catalog No. S19436-4
Description
Description
  • Show the three-dimensional lattice structure of each crystal
  • Visualize and explain properties of substances, concept of unit cells, and interatomic symmetry
  • Different colored spheres are used to represent crystal structures containing more than one type of atom
  • Diamond model is the most complex with 84 spheres
Specifications
Specifications
3-D lattice structure of crystal
Salt
Crystal molecular model
Documents