Please login to your online account to display your discounted pricing

Crystal Formation Lab Investigation


Grow beautiful crystals in just days.

Manufacturer: fisher science education  60-5260

 View more versions of this product

Catalog No. S01419

  • / Each
 

Description & Specifications

Specifications

Grade Level 7 to 10
Product Type Growth Paper
Class Size 40 students
For Use With (Application) For crystal formation lab investigation