Please login to your online account to display your discounted pricing

Crystal Formation Lab Investigation

Grow beautiful crystals in just days.

$17.90 - $17.90

Specifications

Class Size 40 students
Grade Level 7 to 10
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Product Type Price Quantity    

S01419

 
fisher science education
60-5260
For crystal formation lab investigation Growth Paper Each for $17.90
Description & Specifications

Specifications

Class Size 40 students
Grade Level 7 to 10

  • Introduce geometric shapes, symmetry, properties of minerals, gemology, and more
  • Special growth papers grow amazing crystals
  • Students examine shapes and build models of each crystal system
  • Class size: 40 students

Grades 7-10

NSES: A, B, E, F, G