missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cryptotanshinone, TCI America™

Manufacturer:  TCI America C3363100MG

 View more versions of this product

Catalog No. C3363100MG



Specifications

Specifications

Cryptotanshinone
192°C
35825-57-1
MFCD07636810
Tanshinone C, (R)-1,2,6,7,8,9-Hexahydro-1,6,6-trimethylphenanthro[1,2-b]furan-10,11-dione
CC1COC2=C1C(=O)C(=O)C3=C2C=CC4=C3CCCC4(C)C
296.366
296.37
≥97.0% (HPLC)
Undesignated
100mg
C19H20O3
1544
GVKKJJOMQCNPGB-JTQLQIEISA-N
(1R)-1,6,6-trimethyl-2,7,8,9-tetrahydro-1H-naphtho[1,2-g][1]benzofuran-10,11-dione
160254
Crystal-Powder at 20°C

Chemical Identifiers

35825-57-1
296.366
GVKKJJOMQCNPGB-JTQLQIEISA-N
160254
CC1COC2=C1C(=O)C(=O)C3=C2C=CC4=C3CCCC4(C)C
C19H20O3
MFCD07636810
Tanshinone C, (R)-1,2,6,7,8,9-Hexahydro-1,6,6-trimethylphenanthro[1,2-b]furan-10,11-dione
(1R)-1,6,6-trimethyl-2,7,8,9-tetrahydro-1H-naphtho[1,2-g][1]benzofuran-10,11-dione
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents