Please login to your online account to display your discounted pricing

Tocris™ Cryptotanshinone

STAT3 inhibitor. Also displays multiple other activities

$164.61 - $682.67

Specifications

Product Type Cryptotanshinone
Inhibitors Cryptotanshinone
Molecular Weight 296.36
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

371310R

 
tocris™
RDS371310
10mg Each for $164.61

371350R

 
tocris™
RDS371350
50mg Each for $682.67
Description & Specifications

Specifications

Product Type Cryptotanshinone
Inhibitors Cryptotanshinone
Molecular Weight 296.36
Purity >98

Major tanshinone isolated from Salvia miltiorrhiza that exhibits multiple activities. Exhibits antitumor activity via inhibition STAT3 activity (IC50 = 4.6μM). Displays antibacterial and anti-inflammatory activity and acts as an antidiabetes and antiobesity agent via activation of AMP-activated protein kinase (AMPK). Also improves cognitive impairment in Alzheimer's disease transgenic mice by inhibition of acetylcholinesterase (IC50 = 4.09μM) and reduction in Aβ peptide generation.

  • Chemical Name: 1,2,6,7,8,9-Hexahydro-1,6,6-trimethyl[1,2-b]furan-10,11-dione
  • Formula: C19H20O3
  • CAS Number: 35825-57-1
  • Solubility: Soluble to 10mM in DMSO
  • Purity: >98