Please login to your online account to display your discounted pricing
Thermo Scientific™ Cryoware Labels
Easily and permanently identify samples and storage boxes with Thermo Scientific™ Cryoware Labels.
$176.55 - $176.55
Products
Catalog Number Mfr. No. Price Quantity    
03-337-7G
thermo scientific™
N50400002
Pack of 200 Each for $176.55
Case of 10 Pack for $1,765.50
Description & Specifications
Easily and permanently identify samples and storage boxes with Thermo Scientific™ Cryoware Labels.
  • 25 x 50mm cloth labels
  • Accept markers or ballpoint pen
  • Specifically designed for use at ultra-low temperatures
  • Adhere to plastic and cardboard without shrinking or peeling
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.