Thermo Scientific™ CryoPlus™ Vapor Phase Starter Packages
Thermo Scientific™ CryoPlus™ vapor phase starter packages include auto-fill LN2 CryoPlus with racks, riser and hot gas bypass.
$31306.00 - $38593.18
Specifications
Type Auto-Fill LN2 Cryo
No. of Doors 1
Interior Stainless Steel
Certifications/Compliance CE, CFDA, cULus
Style Upright
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
13-987-213
View Documents Promotion Details
Thermo Scientific™
7400VP
90 L N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-987-214
View Documents Promotion Details
Thermo Scientific™
7402VP
200 L N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Thermo Scientific CryoPlus starter packages include the equipment needed for vapor phase, liquid nitrogen storage. All packages include a CryoPlus auto-fill LN2 system with racks, riser and hot gas bypass.
  • Choose 90L CryoPlus 1 or 200L CryoPlus 2
  • 2mL vial capacities: 6,318 (CryoPlus 1), 13,500 (CryoPlus 2)
  • Top-mounted control panel allow easy access to the microprocessor controller for programming
  • Security features: front-mounted key lock, microprocessor key lock and rear-mounted recessed power switch to prevent accidental power shut-off
  • 24 tricolor LEDs continually display actual liquid nitrogen level and high-level/low-level setpoints
  • Counterbalanced lid with 100% clearance allows each entry into the storage area for retrieval of samples while minimizing exposure time
  • Temperature sleeve incorporated, providing colder temperatures at the top and a more efficient vapor phase operation
  • Vacuum insulated with stainless steel interior
  • LN2 transfer hose included 
Specifications
Specifications
Auto-Fill LN2 Cryo
Stainless Steel
Upright
CryoPlus
NEMA 5-15
60 Hz
1
CE, CFDA, cULus
Microprocessor, LED
Cabinet and Microprocessor Key Lock
115 V
SDS
missing translation for 'documents'
missing translation for 'documents'