Please login to your online account to display your discounted pricing

Crotyl Chloride (Tech.), 70%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  154805000

 View more versions of this product

Catalog No. AC154805000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥65%
Purity Grade Technical Grade
Formula Weight 90.55
Physical Form Liquid
Quantity 500mL
Color Clear colorless
Merck Index 15, 2130
Beilstein 01, II, 176
RTECS Number EM4264000
EINECS Number 209-739-5
TSCA TSCA
CAS 563-52-0
Linear Formula CH3CH=CHCH2Cl
CAS 591-97-9
Name Note tech., 70%
Chemical Name or Material Crotyl chloride
Assay Percent Range 70%
MDL Number MFCD00000987
Molecular Formula C4H7Cl
Refractive Index 1.4300 to 1.4360 (20°C, 589nm)
Packaging Glass bottle
Synonym 1-Chloro-2-butene
CAS 1190-22-3
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Crotyl chloride
Name Note tech., 70%
CAS 591-97-9
CAS 563-52-0
CAS 1190-22-3
Purity Grade Technical Grade
Assay ≥65%
Assay Percent Range 70%
Molecular Formula C4H7Cl
Linear Formula CH3CH=CHCH2Cl
Formula Weight 90.55
MDL Number MFCD00000987
Synonym 1-Chloro-2-butene
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

SDS & Safety Documents