missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Crotyl chloride, 70%, tech., Acros Organics

Manufacturer:  Thermo Fisher Scientific 154801000

 View more versions of this product

Catalog No. AC154801000



Specifications

Specifications

Crotyl chloride
−15°C
Glass bottle
0.92
80.0°C to 85.0°C
-65.0°C
tech., 70%
65% min. (GC)
CH3CH=CHCH2Cl
01, II, 176
1-Chloro-2-butene
90.55
Liquid
Technical
0.9200g/mL
Authentic
1.4300 to 1.4360
0.46 mPa.s (20°C)
Colorless
100mL
591-97-9,563-52-0,1190-22-3
C4H7Cl
MFCD00000987
15, 2130
Solubility in water:+/- 14g/L (20°C)
90.55
70%
Documents