missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Crotonoyl Chloride (cis- and trans- mixture) 95.0+%, TCI America™

Supplier:  TCI America C0424100G

 View more versions of this product

Catalog No. C0424100G



Description

Description

Specifications

Specifications

Crotonoyl Chloride (cis- and trans- mixture)
Yellow
C4H5ClO
100 g
RJUIDDKTATZJFE-NSCUHMNNSA-N
(E)-but-2-enoyl chloride
643131
≥95.0% (GC)
10487-71-5
125°C
MFCD00000734
2920
CC=CC(=O)Cl
104.533
104.53
Liquid

Chemical Identifiers

10487-71-5
104.533
RJUIDDKTATZJFE-NSCUHMNNSA-N
(E)-but-2-enoyl chloride
C4H5ClO
MFCD00000734
643131
CC=CC(=O)Cl
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3377

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions