missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cromolyn sodium salt, 98%, Thermo Scientific Chemicals

Mast cell stabilizer that prevents release of histamine from mast cells

Supplier:  Thermo Scientific Chemicals J6512206

 View more versions of this product

Catalog No. AAJ6512206



Description

Description

Cromolyn sodium salt is used as a mast cell stabilizer that prevents release of histamine and other pro-inflammatory mediators from mast cells. Cromolyn sodium salt is used as a chemopreventive and an anti-inflammatory agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cromolyn sodium salt is used as a mast cell stabilizer that prevents release of histamine and other pro-inflammatory mediators from mast cells. Cromolyn sodium salt is used as a chemopreventive and an anti-inflammatory agent.

Solubility
Soluble in water to 100mg/ml

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Cromolyn sodium salt
White to Yellow
MFCD00057744
14,2590
Soluble in water to 100mg/ml
[Na+].[Na+].OC(COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O)COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O
512.33
CHEBI:128458
98%
15826-37-6
C23H14Na2O11
5 g
cromolyn sodium, sodium cromoglycate, disodium cromoglycate, sodium cromoglicate, sodium cromolyn, gastrocrom, nasalcrom, cromoglycate disodium, natrium cromoglicat, lomudal
VLARUOGDXDTHEH-UHFFFAOYSA-L
disodium;5-[3-(2-carboxylato-4-oxochromen-5-yl)oxy-2-hydroxypropoxy]-4-oxochromene-2-carboxylate
27503
512.33
Powder

Chemical Identifiers

15826-37-6
512.33
VLARUOGDXDTHEH-UHFFFAOYSA-L
27503
[Na+].[Na+].OC(COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O)COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O
C23H14Na2O11
MFCD00057744
cromolyn sodium, sodium cromoglycate, disodium cromoglycate, sodium cromoglicate, sodium cromolyn, gastrocrom, nasalcrom, cromoglycate disodium, natrium cromoglicat, lomudal
CHEBI:128458
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 239-926-7

RTECSNumber : DJ2380000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only