Please login to your online account to display your discounted pricing

Cresol Red Sodium Salt, Reagent Grade, LabChem™

$94.83 - $94.83

Chemical Identifiers

Chemical Name or Material Cresol Red Sodium Salt
CAS 62625-29-0
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C21H17O5SNa
Formula Weight 404.42
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC135107

 
lab chem
LC135107
10g Each for $94.83
Description & Specifications

Specifications

Formula Weight 404.42
Packaging Amber Glass
Color Brown
CAS 62625-29-0
Physical Form Powder
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide
Identification Passes Test
Linear Formula C21H17O5SNa
Quantity 10g
Health Hazard 2 GHS H Statement
Solution is not hazardous.
EINECS Number 263-654-8
CAS Max % 100
Purity Grade Reagent Grade
Chemical Name or Material Cresol Red Sodium Salt
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Melting Point 250°C
Recommended Storage Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature