missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cresol Red, 0.04% Aqueous, pH 0.2 to 1.8 Red to Yellow, pH 7.2 to 8.8 Yellow to Purple, Certified, LabChem™
Supplier: LabChem LC135207
Specifications
Cresol Red | |
62625-29-0,7732-18-5 | |
C21H17O5SNa | |
Passes Test | |
Certified | |
1g/mL | |
1g/mL | |
125 mL |
0.04% Aqueous, pH 0.2 to 1.8 Red to Yellow, pH 7.2 to 8.8 Yellow to Purple | |
99.96,0.04 | |
Soluble in water | |
404.42 | |
Poly Bottle | |
Sulfur compounds; Carbon monoxide; Carbon dioxide | |
Red | |
Liquid |
Chemical Identifiers
62625-29-0 |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature