missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Creatine phosphate disodium salt tetrahydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 337340100

 View more versions of this product

Catalog No. AC337340100



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Creatine phosphate disodium salt tetrahydrate
White
99%
C4H8N3Na2O5P
MFCD00150192
creatine phosphate sodium salt
RNTXMYSPASRLFT-UHFFFAOYSA-L
disodium tetrahydrate ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
91884830
99%
71519-72-7
Authentic
Glass bottle
Na2O3PNHC(NH)N(CH3)CH2COOH·4H2O
10 g
(10% in water) Clear colorless solution
[Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
255.08
327.14
Powder

Chemical Identifiers

71519-72-7
255.08
RNTXMYSPASRLFT-UHFFFAOYSA-L
91884830
C4H8N3Na2O5P
MFCD00150192
creatine phosphate sodium salt
[Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only