missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Creatine monohydrate, 99%

$54.65 - $671.81

Chemical Identifiers

CAS 6020-87-7
Molecular Formula C4H11N3O3
Molecular Weight (g/mol) 149.15
MDL Number MFCD00071582
InChI Key MEJYXFHCRXAUIL-UHFFFAOYSA-N
Synonym 2-1-methylguanidino acetic acid hydrate, creapure, creatine hydrate, creatine monohydrate, creatine, monohydrate, creatinemonohydrate, glycine, n-aminoiminomethyl-n-methyl-, monohydrate, n-amidinosarcosine monohydrate, n-aminoiminomethyl-n-methylglycine monohydrate, unii-9603ln7r2q
PubChem CID 80116
IUPAC Name 2-[carbamimidoyl(methyl)amino]acetic acid;hydrate
SMILES CN(CC(=O)O)C(=N)N.O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2500922
View Documents
Alfa Aesar™
B2500922
100g Each for $54.65
 
AAB2500936
View Documents
Alfa Aesar™
B2500936
500g Each for $178.41
 
AAB250090E
View Documents
Alfa Aesar™
B250090E
2500g Each for $671.81
 
Description

Description

Solubility
Soluble in water (17 g/L) at 20°C. Insoluble in ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications

Chemical Identifiers

6020-87-7
149.15
MEJYXFHCRXAUIL-UHFFFAOYSA-N
80116
CN(CC(=O)O)C(=N)N.O
C4H11N3O3
MFCD00071582
2-1-methylguanidino acetic acid hydrate, creapure, creatine hydrate, creatine monohydrate, creatine, monohydrate, creatinemonohydrate, glycine, n-aminoiminomethyl-n-methyl-, monohydrate, n-amidinosarcosine monohydrate, n-aminoiminomethyl-n-methylglycine monohydrate, unii-9603ln7r2q
2-[carbamimidoyl(methyl)amino]acetic acid;hydrate

Specifications

Weak
100g
6020-87-7
MFCD00071582
14,2568
Soluble in water (17g/L) at 20°C. Insoluble in ether.
CN(CC(=O)O)C(=N)N.O
149.15
149.15 (131.13 Anhydrous)
Creatine monohydrate
~292°C (decomposition)
6.9
C4H11N3O3
907175
2-1-methylguanidino acetic acid hydrate, creapure, creatine hydrate, creatine monohydrate, creatine, monohydrate, creatinemonohydrate, glycine, n-aminoiminomethyl-n-methyl-, monohydrate, n-amidinosarcosine monohydrate, n-aminoiminomethyl-n-methylglycine monohydrate, unii-9603ln7r2q
MEJYXFHCRXAUIL-UHFFFAOYSA-N
2-[carbamimidoyl(methyl)amino]acetic acid;hydrate
80116
99%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 200-306-6

RTECSNumber : MB7706000

TSCA : Yes

Recommended Storage : Ambient temperatures