Mopec™ Creased Dissection Table without Drawers
Designed for the dissection of cadavers and offers a broad dissection area. Mopec™ Creased Dissection Table without Drawers promotes easier transfer of bodies on and off the table.
Supplier:  Mopec™ HA210DUP
Catalog No. FSTP9778941
Description
Description
  • All stainless steel construction
  • Creased non-removable top
  • Built-in tapered slope
  • Includes drain hole PVC drain valve
  • Four heavy duty 5 in. casters with 3 total locks and 1 directional lock
  • 4 Post sockets included
Warranty
1 Year Warranty