missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Crayola™ Colored Pencils

Assorted colored pencils designed for classroom and art room activities make it easy to lay down color and blend without a scratch.

$11.15 - $11.15

Specifications

Certifications ASTM D-4236
Color Assorted
For Use With (Application) For classroom and art room activities
Thickness 0.33cm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S98324 Fisher Science Education
550-996
24 Pack Pack of 24 Each for $11.15
Specifications

Description

  • Thick 0.13 in. (3.3mm) soft leads
  • Pre-sharpened pencils facilitate use
  • Not washable

Certifications

Certified AP nontoxic by the Art and Craft Materials Institute and conform to ASTM™ D-4236.

Description & Specifications

Specifications

ASTM D-4236
Assorted
For classroom and art room activities
0.33cm