Please login to your online account to display your discounted pricing

Crayola™ Colored Pencils


Assorted colored pencils designed for classroom and art room activities make it easy to lay down color and blend without a scratch.

Manufacturer: fisher science education  550-996

 View more versions of this product

Catalog No. S98324

  • / Pack of 24 Each
 

Description & Specifications

Specifications

Certifications ASTM D-4236
Product Type Crayola™ Color Pencil
Color Assorted
For Use With (Application) For classroom and art room activities
Quantity 24 Pack
Thickness 0.33cm