missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ CR3600 CHARGING STATION W/ EMBEDDED MODEM and 3 ft. USB CHARGE CABLE
Power supply formatted for North America
Supplier: Brady™ 143571
Description
With embedded modem and 3 ft. USB charge cableSpecifications
Battery Charging Station | |
242.57 mm |