Please login to your online account to display your discounted pricing

Micronova™ CR100 PC™ Cleanroom Tape—Dark Blue

Processed for applications in and around cleanrooms

$22.65 - $22.65

Specifications

Color Dark Blue
Format Tape
Product Type Cleanroom Tape
Quantity Each
Sterile No
View More Specs
Products
Catalog Number Mfr. No. Width (English) Price Quantity    

29-636-4344

 
micronova™
CR100PC34DB
0.75 in. Each for $22.65
Description & Specifications

Specifications

Color Dark Blue
Format Tape
Product Type Cleanroom Tape
Quantity Each
Sterile No
Finish Matte
For Use With (Application) Sealing, color coding, labeling, batching
Length (English) 36 yd.
Length (Metric) 33m
Material Vinyl with Synthetic Rubber

  • Made of vinyl with synthetic rubber adhesive that ensures clean peel from wafer cartridges and most cleanroom work surfaces
  • Tapes and seals curved and irregular surfaces
  • Matte finish can be written on with cleanroom pen
  • Individually packaged in polyethylene bag

Available in red, yellow, white, black, brown, dark blue, dark green, light blue, light green, orange, purple, and transparent in 36 yd. long (33m) rolls. Contact your Fisher Safety Customer Service Representative at 1-800-772-6733 for ordering information on colors not shown here.