Please login to your online account to display your discounted pricing

CPPG, Tocris


Very potent group III mGlu antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS097250

 View more versions of this product

Catalog No. 097250

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Physical Form Solid
Formula Weight 271.21
Chemical Name or Material CPPG
Molecular Formula C11H14NO5P
CAS 183364-82-1
Synonym (RS)-α-Cyclopropyl-4-phosphonophenylglycine
Assay Percent Range >98%
Recommended Storage Store at Room Temperature

Specifications

Physical Form Solid
Accessories

Store at Room Temperature