Please login to your online account to display your discounted pricing

CPPG, Tocris

Very potent group III mGlu antagonist

$214.12 - $895.76

Chemical Identifiers

Physical Form Solid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

097250

 
r & d systems
RDS097250
50mg Each for $895.76

097210

 
r & d systems
RDS097210
10mg Each for $214.12
Description & Specifications

Specifications

Quantity 50mg
Physical Form Solid
Formula Weight 271.21
Synonym (RS)-α-Cyclopropyl-4-phosphonophenylglycine
Molecular Formula C11H14NO5P
CAS 183364-82-1
Chemical Name or Material CPPG
Assay Percent Range >98%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature