Please login to your online account to display your discounted pricing

CPCCOEt, Tocris


Selective non-competitive mGlu1 receptor antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS102810

 View more versions of this product

Catalog No. 102810

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 247.25
Quantity 10mg
Assay Percent Range >99%
Molecular Formula C13H13NO4
CAS 179067-99-3
Synonym 7-(Hydroxyimino)cyclopropa[b]chromen-1a-carboxylate ethyl ester
Chemical Name or Material CPCCOEt
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material CPCCOEt
Assay Percent Range >99%
Molecular Formula C13H13NO4
Formula Weight 247.25
Synonym 7-(Hydroxyimino)cyclopropa[b]chromen-1a-carboxylate ethyl ester
Accessories

Store at Room Temperature