Please login to your online account to display your discounted pricing

CP 99994 dihydrochloride, Tocris™

High affinity NK1 antagonist

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material CP 99994 dihydrochloride
Assay Percent Range >99%
Molecular Formula C19H24N2O·2HCl
Formula Weight 369.33
Synonym (2S, 3S)-N-[(2-Methoxyphenyl)methyl]-2-phenyl-3-piperidinamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

341750

 
r & d systems
RDS341750
50mg Each for $1,180.32

341710

 
r & d systems
RDS341710
10mg Each for $284.81
Description & Specifications

Specifications

Chemical Name or Material CP 99994 dihydrochloride
CAS 145148-39-6
Recommended Storage Desiccate at Room Temperature
Assay Percent Range >99%
Molecular Formula C19H24N2O·2HCl
Formula Weight 369.33
Synonym (2S, 3S)-N-[(2-Methoxyphenyl)methyl]-2-phenyl-3-piperidinamine dihydrochloride
Quantity 50mg
Safety and Handling

Desiccate at Room Temperature