Please login to your online account to display your discounted pricing

CP 945598 hydrochloride, Tocris™

CB1 receptor antagonist

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material CP 945598 hydrochloride
Assay Percent Range >99%
Molecular Formula C25H25Cl2N7O·HCl
Formula Weight 546.88
Synonym 1-[8-(2-Chlorophenyl)-9-(4-chlorophenyl)-9H-purin-6-yl]-4-(ethylamino)piperidine-4-carboxamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

423610

 
r & d systems
423610
10mg Each for $284.81

423650

 
r & d systems
423650
50mg Each for $1,180.32
Description & Specifications

Specifications

Chemical Name or Material CP 945598 hydrochloride
Quantity 10mg
CAS 686347-12-6
Recommended Storage Store at +4°C
Assay Percent Range >99%
Molecular Formula C25H25Cl2N7O·HCl
Formula Weight 546.88
Synonym 1-[8-(2-Chlorophenyl)-9-(4-chlorophenyl)-9H-purin-6-yl]-4-(ethylamino)piperidine-4-carboxamide hydrochloride
Safety and Handling

Store at +4°C