Please login to your online account to display your discounted pricing

CP 94253 hydrochloride, Tocris

Potent, selective 5-HT1B agonist

$233.76 - $930.21

Chemical Identifiers

Chemical Name or Material CP 94253 hydrochloride
Assay Percent Range >98%
Molecular Formula C15H19N3O·HCl
Formula Weight 293.8
Synonym 5-Propoxy-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-pyrrolo[3,2-b]pyridine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

131710

 
r & d systems
RDS131710
10mg Each for $233.76

131750

 
r & d systems
RDS131750
50mg Each for $930.21
Description & Specifications

Specifications

Chemical Name or Material CP 94253 hydrochloride
CAS 845861-39-4
Recommended Storage Desiccate at Room Temperature
Assay Percent Range >98%
Molecular Formula C15H19N3O·HCl
Formula Weight 293.8
Synonym 5-Propoxy-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-pyrrolo[3,2-b]pyridine hydrochloride
Quantity 10mg
Safety and Handling

Desiccate at Room Temperature