CP 94253 hydrochloride, Tocris
Potent, selective 5-HT1B agonist
Manufacturer:  R & D Systems RDS131710
 View more versions of this product
Catalog No. 131710
Specifications Description & Specifications
Specifications
CP 94253 hydrochloride
>98%
293.8
10mg
845861-39-4
C15H19N3O·HCl
5-Propoxy-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-pyrrolo[3,2-b]pyridine hydrochloride
Chemical Identifiers
>98%
293.8
CP 94253 hydrochloride
C15H19N3O·HCl
5-Propoxy-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-pyrrolo[3,2-b]pyridine hydrochloride
Safety and Handling
Recommended Storage : Desiccate at Room Temperature