Please login to your online account to display your discounted pricing

CP 93129 dihydrochloride, Tocris

5-HT1B agonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material CP 93129 dihydrochloride
Assay Percent Range >98%
Molecular Formula C12H13N3O·2HCl
Formula Weight 288.18
Synonym 1,4-Dihydro-3-(1,2,3,6-tetrahydro-4-pyridinyl)-5H-pyrrol[3,2-b]pyridin-5-one dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

103250

 
r & d systems
RDS103250
50mg Each for $844.73

103210

 
r & d systems
RDS103210
10mg Each for $201.36
Description & Specifications

Specifications

Formula Weight 288.18
Quantity 50mg
Molecular Formula C12H13N3O·2HCl
Synonym 1,4-Dihydro-3-(1,2,3,6-tetrahydro-4-pyridinyl)-5H-pyrrol[3,2-b]pyridin-5-one dihydrochloride
CAS 879089-64-2
Chemical Name or Material CP 93129 dihydrochloride
Assay Percent Range >98%
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature