Please login to your online account to display your discounted pricing

CP 809101 hydrochloride, Tocris

Potent and selective 5-HT2C agonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material CP 809101 hydrochloride
Assay Percent Range >99%
Molecular Formula C15H17N4OCl·HCl
Formula Weight 341.24
Synonym 2-[(3-Chlorophenyl)methoxy]-6-(1-piperazinyl)pyrazine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

304110

 
r & d systems
304110
10mg Each for $177.37

304150

 
r & d systems
304150
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 341.24
Quantity 10mg
Chemical Name or Material CP 809101 hydrochloride
Synonym 2-[(3-Chlorophenyl)methoxy]-6-(1-piperazinyl)pyrazine hydrochloride
Molecular Formula C15H17N4OCl·HCl
Assay Percent Range >99%
CAS 1215721-40-6
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature