Please login to your online account to display your discounted pricing

CP 80633, Tocris

Selective PDE4 inhibitor

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material CP 80633
Assay Percent Range >99%
Molecular Formula C18H24N2O3
Formula Weight 316.39
Synonym 5-[3-[(1S, 2S, 4R)-Bicyclo[2.2.1]hept-2-yloxy]-4-methoxyphenyl]tetrahydro-2(1H)-pyrimidinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

272910

 
r & d systems
272910
10mg Each for $244.81

272950

 
r & d systems
272950
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 316.39
Quantity 10mg
Assay Percent Range >99%
CAS 135637-46-6
Molecular Formula C18H24N2O3
Recommended Storage Store at +4°C
Chemical Name or Material CP 80633
Synonym 5-[3-[(1S, 2S, 4R)-Bicyclo[2.2.1]hept-2-yloxy]-4-methoxyphenyl]tetrahydro-2(1H)-pyrimidinone
Safety and Handling

Store at +4°C