Please login to your online account to display your discounted pricing

CP 775146, Tocris™

Selective, high affinity PPARα agonist

$290.95 - $1,205.80

Chemical Identifiers

Chemical Name or Material CP 775146
Assay Percent Range >99%
Molecular Formula C26H33NO4
Formula Weight 423.54
Synonym 2-Methyl-2-[3-[(3S)-1-[2-[4-(1-methylethyl)phenyl]acetyl]-3-piperidinyl]phenoxy]-propanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

419050

 
r & d systems
RDS419050
50mg Each for $1,205.80

419010

 
r & d systems
RDS419010
10mg Each for $290.95
Description & Specifications

Specifications

Formula Weight 423.54
Quantity 50mg
Chemical Name or Material CP 775146
Assay Percent Range >99%
Molecular Formula C26H33NO4
CAS 702680-17-9
Synonym 2-Methyl-2-[3-[(3S)-1-[2-[4-(1-methylethyl)phenyl]acetyl]-3-piperidinyl]phenoxy]-propanoic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature