Please login to your online account to display your discounted pricing

CP 690550 citrate, Tocris™

Potent JAK inhibitor

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material CP 690550 citrate
Assay Percent Range >99%
Molecular Formula C16H20N6O·C6H8O7
Formula Weight 504.49
Synonym (3R, 4R)-4-Methyl-3-(methyl-7H-pyrrolo[2, 3-d]pyrimidin-4-ylamino)-β-oxo-1-piperidinepropanenitrile citrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

455610

 
r & d systems
RDS455610
10mg Each for $214.12

455650

 
r & d systems
RDS455650
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 504.49
Quantity 10mg
Chemical Name or Material CP 690550 citrate
Assay Percent Range >99%
Molecular Formula C16H20N6O·C6H8O7
Synonym (3R, 4R)-4-Methyl-3-(methyl-7H-pyrrolo[2, 3-d]pyrimidin-4-ylamino)-β-oxo-1-piperidinepropanenitrile citrate
CAS 540737-29-9
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C