Please login to your online account to display your discounted pricing

CP 471474, Tocris™

Broad spectrum MMP inhibitor

$192.67 - $812.12

Chemical Identifiers

Chemical Name or Material CP 471474
Assay Percent Range >97%
Molecular Formula C16H17FN2O5S
Formula Weight 368.38
Synonym 2-[[[4-(4-Fluorophenoxy)phenyl]sulfonyl]amino]-N-hydroxy-2-methylpropanamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

378010

 
r & d systems
378010
10mg Each for $192.67

378050

 
r & d systems
378050
50mg Each for $812.12
Description & Specifications

Specifications

Formula Weight 368.38
Quantity 10mg
Chemical Name or Material CP 471474
Molecular Formula C16H17FN2O5S
CAS 210755-45-6
Synonym 2-[[[4-(4-Fluorophenoxy)phenyl]sulfonyl]amino]-N-hydroxy-2-methylpropanamide
Recommended Storage Store at -20°C
Assay Percent Range >97%
Safety and Handling

Store at -20°C