Please login to your online account to display your discounted pricing

CP 465022 hydrochloride, Tocris

Selective, non-competitive AMPA antagonist

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material CP 465022 hydrochloride
Assay Percent Range >99%
Molecular Formula C26H24ClFN4O·HCl
Formula Weight 499.41
Synonym 3-(2-Chlorophenyl)-2-[2-[6-[(diethylamino)methyl]-2-pyridinyl]ethenyl]-6-fluoro-4(3H)-quinazolinone hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

293250

 
r & d systems
293250
50mg Each for $862.95

293210

 
r & d systems
293210
10mg Each for $205.70
Description & Specifications

Specifications

Formula Weight 499.41
Quantity 50mg
Assay Percent Range >99%
Synonym 3-(2-Chlorophenyl)-2-[2-[6-[(diethylamino)methyl]-2-pyridinyl]ethenyl]-6-fluoro-4(3H)-quinazolinone hydrochloride
Chemical Name or Material CP 465022 hydrochloride
Molecular Formula C26H24ClFN4O·HCl
CAS 199655-36-2
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature