Please login to your online account to display your discounted pricing

CP 339818 hydrochloride, Tocris

Non-peptide, potent KV1.3 channel blocker

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material CP 339818 hydrochloride
Assay Percent Range >99%
Molecular Formula C21H24N2·HCl
Formula Weight 340.89
Synonym N-[1-(Phenylmethyl)-4(1H)-quinolinylidene]-1 pentanamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

139910

 
r & d systems
139910
10mg Each for $214.12

139950

 
r & d systems
139950
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 340.89
Quantity 10mg
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Synonym N-[1-(Phenylmethyl)-4(1H)-quinolinylidene]-1 pentanamine hydrochloride
Chemical Name or Material CP 339818 hydrochloride
Molecular Formula C21H24N2·HCl
CAS 478341-55-8
Safety and Handling

Store at Room Temperature