Please login to your online account to display your discounted pricing

CP 316819, Tocris™

Selective glycogen phosphorylase inhibitor

$259.29 - $1,075.94

Chemical Identifiers

Chemical Name or Material CP 316819
Assay Percent Range >98%
Molecular Formula C21H22ClN3O4
Formula Weight 415.87
Synonym 5-Chloro-N-[(1S, 2R)-2-hydroxy-3-(methoxymethylamino)-3-oxo-1-(phenylmethyl)propyl]-1H-indole-2-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

354210

 
r & d systems
354210
10mg Each for $259.29

354250

 
r & d systems
354250
50mg Each for $1,075.94
Description & Specifications

Specifications

Chemical Name or Material CP 316819
Quantity 10mg
CAS 186392-43-8
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C21H22ClN3O4
Formula Weight 415.87
Synonym 5-Chloro-N-[(1S, 2R)-2-hydroxy-3-(methoxymethylamino)-3-oxo-1-(phenylmethyl)propyl]-1H-indole-2-carboxamide
Safety and Handling

Store at Room Temperature