Please login to your online account to display your discounted pricing

CP 31398 dihydrochloride, Tocris

p53-stabilizing agent

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material CP 31398 dihydrochloride
Assay Percent Range >99%
Molecular Formula C22H26N4O·2HCl
Formula Weight 435.39
Synonym N'-[2-[2-(4-Methoxyphenyl)ethenyl]-4-quinazolinyl]-N, N-dimethyl-1, 3-propanediamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

302350

 
r & d systems
302350
50mg Each for $844.73

302310

 
r & d systems
302310
10mg Each for $201.36
Description & Specifications

Specifications

Chemical Name or Material CP 31398 dihydrochloride
Quantity 50mg
CAS 1217195-61-3
Recommended Storage Desiccate at Room Temperature
Assay Percent Range >99%
Molecular Formula C22H26N4O·2HCl
Formula Weight 435.39
Synonym N'-[2-[2-(4-Methoxyphenyl)ethenyl]-4-quinazolinyl]-N, N-dimethyl-1, 3-propanediamine dihydrochloride
Safety and Handling

Desiccate at Room Temperature