Please login to your online account to display your discounted pricing

CP 20961, Tocris™

Non-immunogenic adjuvant

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material CP 20961
Assay Percent Range >98%
Molecular Formula C43H90N2O2
Formula Weight 667.19
Synonym N,N-Dioctadecyl-N',N'-bis(2-hydroxyethyl)-1,3-diaminopropane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

376710

 
r & d systems
RDS376710
10mg Each for $214.12

376750

 
r & d systems
RDS376750
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 667.19
Quantity 10mg
Chemical Name or Material CP 20961
Assay Percent Range >98%
Molecular Formula C43H90N2O2
Synonym N,N-Dioctadecyl-N',N'-bis(2-hydroxyethyl)-1,3-diaminopropane
CAS 35607-20-6
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature